Merck
  • Selective and sentivive method based on capillary liquid chromatography with in-tube solid phase microextraction for determination of monochloramine in water.
Selective and sentivive method based on capillary liquid chromatography with in-tube solid phase microextraction for determination of monochloramine in water.
Journal of chromatography. A (2015-03-03)
J Pla-Tolós, Y Moliner-Martínez, C Molins-Legua, R Herráez-Hernández, J Verdú-Andrés, P Campíns-Falcó
ZUSAMMENFASSUNG
Due to the difficulties of working with chloramines, a critical examination of monochloramine standard preparation has been performed in order to select the best synthesis conditions. The analyte has been determined by in-tube solid phase extraction coupled to capillary liquid chromatography with UV detection (IT-SPME Capillary LC DAD). Potential factors affecting the response of monochloramine such as the pH of mobile phase and the volume of sample processed by IT-SPME Capillary LC DAD have been investigated and optimized. According to the results of the study, 0.1 mL or 4.0 mL of sample at neutral pH were loaded in the chromatographic system. A sensitive and selective method has been developed for the determination of monochloramine in water. Validation of the method has been performed. The linear range was 0.09-5mg/L with linear regression coefficients (R(2)) greater than 0.995. Method reproducibility expressed as relative standard deviation (RSD, %), was lower than 15%. The limits of detection (LODs) were 0.029 and 0.01 mg/L by processing 0.100mL or 4 mL of the samples, respectively, being below the maximum residues levels allowed for this compound. The sensitivity achieved by the developed method was better than that obtained by the reference method. The developed method was applied to water samples (tap and swimming pool water).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumchlorid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Ammoniumchlorid, 99.998% trace metals basis
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in dioxane
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chlor, ≥99.5%
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in isopropanol
Sigma-Aldrich
Thymol, FCC, FG
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub