Merck
  • T-cadherin association with clinicopathological features and prognosis in axillary lymph node-positive breast cancer.
T-cadherin association with clinicopathological features and prognosis in axillary lymph node-positive breast cancer.
Breast cancer research and treatment (2015-02-14)
De-di Kong, Jie Yang, Liang Li, Wei Wang, Ya-Ning Chen, Shi-Bing Wang, Yan-Zhen Zhou
ZUSAMMENFASSUNG
The purpose of this study was to investigate the correlation of T-cadherin expression with clinicopathological features and prognosis in patients with axillary lymph node-positive breast cancer. Based on the immunohistochemistry results, all 142 patients with operable axillary lymph node-positive breast cancer were divided into the T-cadherin-negative and T-cadherin-positive groups. Clinical data including the association of T-cadherin expression with clinicopathological features and prognosis were analyzed using the Chi square test and Fisher's exact test using SPSS 13.0 software. The impact of T-cadherin expression on the 5-year disease-free survival (DFS) and the 5-year overall survival (OS) of these patients was measured using the log-rank test. DFS and OS were analyzed using both Kaplan-Meier function and Cox regression analyses. Compared with the T-cadherin-positive group (55.07, 28.99, and 13.4 %, respectively; P = 0.030, P = 0.0132, and P = 0.009), tumor size >2 cm, lymph-vascular invasion, and pathological stage III disease were seen more frequently in the T-cadherin-negative group (72.60, 49.32, and 31.51 %, respectively). Both 5-year DFS and 5-year OS were poorer in the T-cadherin-negative group than in the T-cadherin-positive group (log-rank test = 9.295, P = 0.002; log-rank test = 5.718, P = 0.017). On multivariate analysis, T-cadherin-negative expression remained an independent prognostic factor for DFS (P = 0.002) but not for OS (P = 0.067). Our results suggested that negative T-cadherin expression has a worse prognosis in patients with axillary lymph node-positive breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesteron, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesteron, United States Pharmacopeia (USP) Reference Standard
Progesteron für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Progesteron, European Pharmacopoeia (EP) Reference Standard
Progesteron für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesteron, VETRANAL®, analytical standard