Merck
MUC1 Promoter-Driven DTA as a Targeted Therapeutic Strategy against Pancreatic Cancer.
Molecular cancer research : MCR (2014-10-23)
Renee M Tholey, Shruti Lal, Masaya Jimbo, Richard A Burkhart, Fernando F Blanco, Joseph A Cozzitorto, Josh D Eisenberg, Wei Jiang, Christine A Iacobuzio-Donahue, Agnieszka K Witkiewicz, Melissa Glbert, Charles J Yeo, Jonathan R Brody, Janet A Sawicki, Jordan M Winter
ZUSAMMENFASSUNG
Mucin1 (MUC1) is overexpressed in pancreatic ductal adenocarcinoma (PDA) and is associated with tumor aggressiveness, suggesting that MUC1 is a promising therapeutic target for promoter-driven diphtheria toxin A (DTA). Endogenous MUC1 transcript levels were analyzed by quantitative PCR (qPCR) in multiple PDA cells (Capan1, HPAFII, Su.86.86, Capan2, Hs766T, MiaPaCa2, and Panc1). Expression levels were correlated with luciferase activity and cell death after transfection with MUC1 promoter-driven luciferase and DTA constructs. MUC1-positive (+) cells had significantly elevated MUC1 mRNA expression compared with MUC1-negative (-) cells. Luciferase activity was significantly higher in MUC1(+) cells when transfected with MUC1 promoter-driven luciferase and MUC1(+) cells underwent enhanced cell death after transfection with a single dose of MUC1 promoter-driven DTA. IFNγ pretreatment enhanced MUC1 expression in MUC1(-) cells and induced sensitivity to MUC1-DTA therapy. Matched primary and metastatic tumor lesions from clinical specimens revealed similar MUC1 IHC labeling patterns, and a tissue microarray of human PDA biopsies revealed increased immunolabeling with a combination of MUC1 and mesothelin (MSLN) antibodies, compared with either antibody alone. Combining MUC1 with MSLN-targeted DTA enhanced drug efficacy in an in vitro model of heterogeneous PDA. These data demonstrate that MUC1 promoter-driven DTA preferentially kills MUC1-expressing PDA cells and drugs that enhance MUC1 expression sensitize PDA cells with low MUC1 expression. MUC1 expression in primary and metastatic lesions provides a rationale for the development of a systemic MUC1 promoter-driven DTA therapy that may be further enhanced by combination with other promoter-driven DTA constructs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trimesinsäure, 95%
SAFC
L-Glutamin
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in toluene
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, suitable for microscopy
Sigma-Aldrich
Trimethylaluminium, 97%
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Trimethylaluminium, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in hexanes
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in heptane
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland