Merck
  • Development of a Matrix Solid Phase Dispersion methodology for the determination of triazine herbicides in mussels.
Development of a Matrix Solid Phase Dispersion methodology for the determination of triazine herbicides in mussels.
Food chemistry (2014-12-04)
N Rodríguez-González, M J González-Castro, E Beceiro-González, S Muniategui-Lorenzo
ZUSAMMENFASSUNG
A method based on Matrix Solid Phase Dispersion (MSPD) for determination of nine triazines in mussels has been optimised in terms of the sorbents used for extracting and cleaning-up. Two dispersing agents: C18 and florisil, and eight cleanup co-sorbents: florisil, silica, silica/alumina, Envi™ Carb, Envi-Carb-II/PSA, SAX/PSA, Envi-Carb-II /SAX/PSA and C18 were assayed. Analytes were eluted using 20 mL of ethyl acetate and 5 mL of acetonitrile and finally the extract was concentrated to dryness, re-constituted with 1 mL methanol and determined by HPLC-DAD. The best results were obtained with C18 as dispersing agent and Envi-Carb-II/SAX/PSA as clean-up co-column. Recoveries ranged between 79% and 99% and repeatability and reproducibility were below than 16% for all compounds. The linearity of the calibration curves yielded the R(2)⩾0.9993. The LOQ values ranged from 0.10 to 0.18 mg kg(-1) dried sample. Finally the method was applied to the analysis of mussel samples from Galicia (NW Spain).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Aluminiumoxid, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Aluminiumoxid, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hexan, anhydrous, 95%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Aluminiumoxid, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminiumoxid, powder, 99.99% trace metals basis
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Hexan, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Atrazin, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminiumoxid, Corundum, α-phase, -100 mesh