Merck
  • Pseudophosphorylation of Tau at distinct epitopes or the presence of the P301L mutation targets the microtubule-associated protein Tau to dendritic spines.
Pseudophosphorylation of Tau at distinct epitopes or the presence of the P301L mutation targets the microtubule-associated protein Tau to dendritic spines.
Biochimica et biophysica acta (2015-01-07)
Di Xia, Chuanzhou Li, Jürgen Götz
ZUSAMMENFASSUNG
Alzheimer's disease is characterized by the accumulation of amyloid-β (Aβ) and Tau in the brain. In mature neurons, Tau is concentrated in the axon and found at lower levels in the dendrite where it is required for targeting Fyn to the spines. Here Fyn mediates Aβ toxicity, which is vastly abrogated when Tau is either deleted or a truncated form of Tau (Tau(1-255)) is co-expressed. Interestingly, MAP2, a microtubule-binding protein with mainly dendritic localization that shares Fyn-binding motifs with Tau, does not mediate Aβ's synaptic toxicity in the absence of Tau. Here we show in hippocampal neurons that endogenous Tau enters the entire spine, albeit at low levels, whereas MAP2 only enters its neck or is restricted to the dendritic shaft. Based on an extensive mutagenesis study, we also reveal that the spine localization of Tau is facilitated by deletion of the microtubule-binding repeat domain. When distinct phosphorylation sites (AT180-T231/S235, 12E8-S262/S356, PHF1-S396/S404) were pseudophosphorylated (with glutamic acid, using alanine replacements as controls), Tau targeting to spines was markedly increased, whereas the pseudophosphorylation of the late phospho-epitope S422 had no effect. In determining the role physiological Fyn has in the spine localization of Tau, we found that neither were endogenous Tau levels reduced in Fyn knockout compared with wild-type synaptosomal brain fractions nor was the spine localization of over-expressed pseudophosphorylated or P301L Tau. This demonstrates that although Fyn targeting to the spine is Tau dependent, elevated levels of phosphorylated Tau or P301L Tau can enter the spine in a Fyn-independent manner.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Supelco
Natriumfluorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumfluorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Natriumfluorid, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumfluorid 0,5 M -Lösung
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
Saccharose, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorid-Ionenlösung für ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar