Merck
MALDI-TOF MS for quality control of high protein content sport supplements.
Food chemistry (2015-01-28)
Cristina De Ceglie, Cosima D Calvano, Carlo G Zambonin
ZUSAMMENFASSUNG
High protein content sport nutritional supplements are found as powder products containing, as ingredients, amino acids and proteins with important nutritional values as milk, soy and egg proteins. An EU Food Supplements Directive (2002) requires that supplements should be safe, both in dosages and in purity. It is important, then, to develop rapid and sensitive methods to be employed for the quality control of these substances. In this work, we apply, for the first time, matrix-assisted laser desorption ionization-mass spectrometry as a fast, reproducible and sensitive method for the quality control of sport nutritional supplements based on proteins. To this aim, several commercial egg- and/or milk-based powder products have been processed by in gel or in solution digestion and analyzed in comparison to pure standard products. This strategy allowed to assess the reliability of the indications on proteins (as caseins, whey proteins and ovalbumin) declared in the label of several sport nutritional supplements.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Ammoniumbicarbonat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
3-Indolessigsäure, 98%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
SAFC
L-Glutamin
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
3-Indolessigsäure, suitable for plant cell culture, crystalline
Sigma-Aldrich
Ammoniumbicarbonat, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%