Merck
  • Determination of polar organic compounds in atmospheric aerosols by gas chromatography with ion trap tandem mass spectrometry.
Determination of polar organic compounds in atmospheric aerosols by gas chromatography with ion trap tandem mass spectrometry.
Journal of separation science (2014-04-12)
Marco Visentin, Maria Chiara Pietrogrande
ZUSAMMENFASSUNG
A gas chromatography with ion trap mass spectrometry method has been developed and validated for the analysis of 27 polar organic compounds in atmospheric aerosols. The target analytes were low-molecular-weight carboxylic acids and methoxyphenols, as relevant markers of source emissions and photochemical processes of organic aerosols. The operative parameters were optimized in order to achieve the best sensitivity and selectivity for the analysis. In comparison with the previous gas chromatography with mass spectrometry procedure based on single ion monitoring detection, the tandem mass spectrometry technique increased the analytical sensitivity by reducing detection limits for standard solutions from 1-2.6 to 0.1-0.4 ng/μL ranges (concentrations in the injected solution). In addition, it enhanced selectivity by reducing matrix interferences and chemical noise in the chromatogram. The applicability of the developed method in air quality monitoring campaigns was effectively checked by analyzing environmental samples collected in the Po Valley (Northern Italy) in different seasons. The obtained results indicate that the ion trap mass spectrometer may be an ideal alternative to high-resolution mass spectrometers for the user-friendly and cost-effective determination of a wide range of molecular tracers in airborne particulate matter.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, derivatization grade (GC derivatization), LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Chlorotrimethylsilan, ≥98.0% (GC)
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoracetamid, ≥99%
Sigma-Aldrich
Chlorotrimethylsilan, purified by redistillation, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoracetamid, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Dodecan, anhydrous, ≥99%
Sigma-Aldrich
Chlorotrimethylsilan, puriss., ≥99.0% (GC)
Sigma-Aldrich
Pyridin, ≥99%
Supelco
Dodecan, analytical standard
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Supelco
Chlorotrimethylsilan, derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, derivatization grade (GC derivatization), LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
Pyridin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridin, analytical standard
Sigma-Aldrich
Chlorotrimethylsilan, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Chlortrimethylsilan -Lösung, 1.0 M in THF
Supelco
Dichte Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality