Merck
Antimicrobial bioassay-guided fractionation of a methanol extract of Eupatorium triplinerve.
Pharmaceutical biology (2014-11-29)
Tamyris Regina Matos Lopes, Fábio Rodrigues de Oliveira, Flávia Filocreão Malheiros, Marcieni Ataíde de Andrade, Marta Chagas Monteiro, Ana Cristina Baetas Gonçalves
ZUSAMMENFASSUNG
Eupatorium triplinerve Vahl (Asteraceae), popularly known as Japana, is widely used in folk medicine, due its analgesic, anticoagulant, antianorexic, antiparasitic, anthelmintic, sedative, antifungal, and antibacterial properties. The present study evaluated the chemical composition and antimicrobial activity of E. triplinerve extracts from different parts of the plant and identified the extract with the highest antimicrobial potential. Extracts were obtained by maceration of all parts of plant, and subjected to bioassay-guided fractionation of methanol extract by partition column chromatography. The major chemical groups, saponins, reducing sugars, alkaloids, steroids, triterpenoids, phenols, tannins, flavonoids, and others were screened by standard techniques. The antimicrobial activity of the different extracts was performed by microdilution assay and the minimum inhibitory concentration (MIC) and minimum bactericidal concentration (MBC) values were reported. Phytochemical screening of hydroalcoholic extract from all parts of E. triplinerve identified mainly steroids, coumarins, alkaloids, saponins, tannins, depsides and absence of polysaccharides and flavonoids. The methanol extract of leaves presented the highest content of coumarins and lower MIC values of 62 and 75 µg/mL against Enterococcus faecalis and Staphylococcus aureus, respectively. In addition, its non-polar fractions showed antimicrobial activity with MIC ranging from 16 to 125 µg/mL against Gram-negative bacteria, mainly Escherichia coli. Data showed that non-polar fractions of E. triplinerve methanolic extract has better antimicrobial activity and most likely depends on the presence of several compounds, such as depsidones, coumarins, saponins, and triterpenes on crude extract. The results can be exploited largely in research of new antibacterial agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.