Merck
  • Reprogramming of mouse somatic cells into pluripotent stem-like cells using a combination of small molecules.
Reprogramming of mouse somatic cells into pluripotent stem-like cells using a combination of small molecules.
Biomaterials (2014-06-03)
Phil Jun Kang, Jai-Hee Moon, Byung Sun Yoon, Solji Hyeon, Eun Kyoung Jun, Gyuman Park, Wonjin Yun, Jiyong Park, Minji Park, Aeree Kim, Kwang Youn Whang, Gou Young Koh, Sejong Oh, Seungkwon You
ZUSAMMENFASSUNG
Somatic cells can be reprogrammed to generate induced pluripotent stem cells (iPSCs) by overexpression of four transcription factors, Oct4, Klf4, Sox2, and c-Myc. However, exogenous expression of pluripotency factors raised concerns for clinical applications. Here, we show that iPS-like cells (iPSLCs) were generated from mouse somatic cells in two steps with small molecule compounds. In the first step, stable intermediate cells were generated from mouse astrocytes by Bmi1. These cells called induced epiblast stem cell (EpiSC)-like cells (iEpiSCLCs) are similar to EpiSCs in terms of expression of specific markers, epigenetic state, and ability to differentiate into three germ layers. In the second step, treatment with MEK/ERK and GSK3 pathway inhibitors in the presence of leukemia inhibitory factor resulted in conversion of iEpiSCLCs into iPSLCs that were similar to mESCs, suggesting that Bmi1 is sufficient to reprogram astrocytes to partially reprogrammed pluripotency. Next, Bmi1 function was replaced with Shh activators (oxysterol and purmorphamine), which demonstrating that combinations of small molecules can compensate for reprogramming factors and are sufficient to directly reprogram mouse somatic cells into iPSLCs. The chemically induced pluripotent stem cell-like cells (ciPSLCs) showed similar gene expression profiles, epigenetic status, and differentiation potentials to mESCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Supelco
Valproinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)