Merck
Rat BMSCs initiate retinal endogenous repair through NGF/TrkA signaling.
Experimental eye research (2015-01-15)
Qian Jian, Yaochen Li, Zheng Qin Yin
ZUSAMMENFASSUNG
Müller cells can completely repair retinal injury by acting as endogenous stem/progenitor cells in lower-order vertebrates. However, a safe and effective approach to activate progenitor potential of retinal Müller cells in higher-order vertebrates, which rarely re-enter the cell cycle, is a bottleneck problem. In the present study, Royal College of Surgeon's (RCS) rats were subjected to rat bone marrow mesenchymal stem cells (rBMSCs) subretinal space transplantation. Electroretinography (ERG) recordings showed that the b-wave amplitudes and ONL thicknesses statistically increased after transplantation. The number of Müller cells expressing proliferative, stem/progenitor and neuronal markers significantly increased after rBMSCs transplantation in vivo or after co-culturing with rBMSCs in vitro. The cultured rBMSCs could secrete nerve growth factor (NGF). In addition, we confirmed that NGF or NGF-neutralizing antibody could activate or depress Müller cells dedifferentiation, both in vivo and in vitro. Furthermore, Müller cells expressing high levels of the NGF receptor neurotrophic tyrosine kinase receptor type 1 (TrkA) were observed in the retinas of rats transplanted with rBMSCs. Moreover, the protein expression of downstream elements of NGF/TrkA signaling, such as p-PI3K, p-Akt and p-CREB, increased in Müller cells in the retinas of rBMSCs-treated rats in vivo or in Müller cells co-cultured with rBMSCs in vitro. Blocking TrkA with K-252a reduced the number of dedifferentiated Müller cells and the expression of NGF/TrkA signaling in vitro. Thus, rBMSCs might initiate endogenous regenerative mechanisms, which may constitute a new therapeutic strategy for retinal dystrophic diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
Saccharose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
K-252a, Ready Made Solution, from Nonomuraea longicatena, >98%