Merck
In vitro bioaccessibility and monolayer uptake of lutein from wholegrain baked foods.
Food chemistry (2014-12-23)
Andrew Read, Amanda Wright, El-Sayed M Abdel-Aal
ZUSAMMENFASSUNG
Lutein-enriched foods would contribute towards promoting the consumption of this important carotenoid. In the current study cookies, muffins, and flatbreads were produced at three enrichment levels (0.5, 1.0 and 2.0mg lutein per serving) to investigate lutein bioavailability in vitro. Foods were subjected to in vitro simulation of human upper gastrointestinal digestion coupled with a Caco-2 monolayer absorption assay. Digestive conditions representative of the fed state conditions associated with significantly higher lutein bioaccessibility compared with the fasted state (p < 0.001). Lutein bioaccessibility of cookies and muffins was higher than flatbreads due to their elevated fat content, measured under either fed or fasted conditions. Lutein concentration in the aqueous phase was the most important factor in determining apical cell uptake of lutein, with cookies and muffins exhibiting a higher lutein uptake than flatbreads (p < 0.05). As commonly consumed foods, lutein-enriched cookies, muffins, and flatbreads have the potential to improve lutein intake.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Lutein, analytical standard
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Zeaxanthin, analytical standard
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C