Merck
  • Composite pullulan-dextran polysaccharide scaffold with interfacial polyelectrolyte complexation fibers: a platform with enhanced cell interaction and spatial distribution.
Composite pullulan-dextran polysaccharide scaffold with interfacial polyelectrolyte complexation fibers: a platform with enhanced cell interaction and spatial distribution.
Acta biomaterialia (2014-07-02)
Marie Francene Arnobit Cutiongco, Ming Hao Tan, Martin Yoke Kuang Ng, Catherine Le Visage, Evelyn King Fai Yim
ZUSAMMENFASSUNG
Hydrogels are highly preferred in soft tissue engineering because they recapitulate the hydrated extracellular matrix. Naturally derived polysaccharides, like pullulan and dextran, are attractive materials with which to form hydrophilic polymeric networks due to their non-immunogenic and non-antigenic properties. However, their inherent hydrophilicity prevents adherent cell growth. In this study, we modified pullulan-dextran scaffolds with interfacial polyelectrolyte complexation (IPC) fibers to improve their ability to support adherent cell growth. We showed that the pullulan-dextran-IPC fiber composite scaffold laden with extracellular matrix protein has improved cell adhesion and proliferation compared to the plain polysaccharide scaffold. We also demonstrated the zero-order release kinetics of the biologics bovine serum albumin and vascular endothelial growth factor (VEGF) incorporated in the composite scaffold. Lastly, we showed that the VEGF released from the composite scaffold retained its capacity to stimulate endothelial cell growth. The incorporation of IPC fibers in the pullulan-dextran hydrogel scaffold improved its functionality and biological activity, thus enhancing its potential in tissue engineering applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Hexamethyldisilazan, reagent grade, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Hexamethyldisilazan, ReagentPlus®, 99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)