Merck
  • Tocotrienols have a nephroprotective action against lipid-induced chronic renal dysfunction in rats.
Tocotrienols have a nephroprotective action against lipid-induced chronic renal dysfunction in rats.
Renal failure (2014-09-19)
Mohammad Rashid Khan, Haseeb Ahsan, Shabeena Siddiqui, Waseem A Siddiqui
ZUSAMMENFASSUNG
Vitamin E is the generic term for a group of tocopherols and tocotrienols (T3). Hyperlipidemia has been known to cause progressive chronic renal dysfunction (CRD). Several investigators have reported that T3 have hypolipidemic and nephroprotective activity against free radical-related diseases. This study was conducted to determine if T3 as tocotrienol-rich fraction (TRF) from palm oil would protect against lipid-induced CRD in rats. For the induction of atherosclerosis and hyperlipidemia, Wistar male rats were fed an atherogenic diet containing 1.25% cholesterol, 0.5% cholic acid and 21% beef tallow (42.6% calories from fat). The atherogenic diet was given for 14 weeks to induce atherosclerosis. The control rats were given normal rat chow and drug control animals treated with TRF (100 mg/kg bw; orally). The first group was taken as disease control in which the animals were left untreated and given normal rat chow for six weeks, while the second group was treated with 100 mg TRF/kg bw. Atherosclerosis and renal functions were evaluated after six weeks of TRF treatment. Feeding an atherogenic diet to rats for 14 weeks resulted in dyslipidemia and impaired renal functions with decreased glomerular filtration rate. The treatment with TRF significantly reduced dyslipidemia and inhibited the development of CRD caused by atherogenic factors. These findings show that low-dose treatment of TRF may provide significant health benefits in the prevention of lipid-induced CRD. The study suggests that TRF is effective in preventing lipid-induced CRD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumnitrit, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumnitrit, ACS reagent, ≥97.0%
Sigma-Aldrich
Natriumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Sulfaminsäure, ACS reagent, 99.3%
Sigma-Aldrich
N-(1-Naphthyl)ethylendiamin -dihydrochlorid, ACS reagent, >98%
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
Sulfaminsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
2,4-Dinitrophenylhydrazin, reagent grade, 97%
Sigma-Aldrich
N-(1-Naphthyl)ethylendiamin -dihydrochlorid, ≥98%
Sigma-Aldrich
Natriumnitrit, 99.999% trace metals basis
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, ≥99%
Sigma-Aldrich
Sulfaminsäure, reagent grade, 98%
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, 97%
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Supelco
Natriumnitrit, analytical standard
Sigma-Aldrich
Natriumnitrat, 99.995% trace metals basis
Supelco
Sulfanilamid Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfaminsäure, 99.999% trace metals basis
Sigma-Aldrich
Thyreotropin-freisetzendes Hormon, ≥98% (HPLC), powder
Supelco
Sulfaminsäure, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
N-(1-Naphthyl)ethylendiamin -dihydrochlorid, ACS reagent, ≥98%
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
2-Nitro-benzoesäure, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Natriumnitrat, ≥99.0%, suitable for plant cell culture
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard