Merck
  • Activation of mu or delta opioid receptors in the lumbosacral spinal cord is essential for ejaculatory reflexes in male rats.
Activation of mu or delta opioid receptors in the lumbosacral spinal cord is essential for ejaculatory reflexes in male rats.
PloS one (2015-04-01)
Natalie Kozyrev, Lique M Coolen
ZUSAMMENFASSUNG
Ejaculation is controlled by a spinal ejaculation generator located in the lumbosacral spinal cord, consisting in male rats of lumbar spinothalamic (LSt) cells and their inter-spinal projections to autonomic and motor centers. LSt cells co-express several neuropeptides, including gastrin releasing peptide (GRP) and enkephalin. We previously demonstrated in rats that GRP regulates ejaculation by acting within the lumbosacral spinal cord. In the present study, the hypothesis was tested that enkephalin controls ejaculation by acting on mu (MOR) or delta opioid receptors (DOR) in LSt target areas. Adult male rats were anesthetized and spinalized and received intrathecal infusions of vehicle, MOR antagonist CTOP (0.4 or 4 nmol), DOR antagonist (TIPP (0.4, 4 or 40 nmol), MOR agonist DAMGO (0.1 or 10 nmol), or DOR agonist deltorphin II (1.3 or 13 nmol). Ejaculatory reflexes were triggered by stimulation of the dorsal penile nerve (DPN) and seminal vesicle pressure and rhythmic contractions of the bulbocavernosus muscle were analyzed. Intrathecal infusion of MOR or DOR antagonists effectively blocked ejaculatory reflexes induced by DPN stimulation. Intrathecal infusion of DAMGO, but not deltorphin II triggered ejaculation in absence of DPN stimulation. Both MOR and DOR agonists facilitated ejaculatory reflexes induced by subthreshold DPN stimulation in all animals. Overall, these results support the hypothesis that enkephalin plays a critical role in the control of ejaculation in male rats. Activation of either MOR or DOR in LSt target areas is required for ejaculation, while MOR activation is sufficient to trigger ejaculation in the absence of sensory stimulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Ethylenglycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethylenglycol, United States Pharmacopeia (USP) Reference Standard
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylenglycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Carbendazim, 97%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)