Merck
  • Lung matrix and vascular remodeling in mechanically ventilated elastin haploinsufficient newborn mice.
Lung matrix and vascular remodeling in mechanically ventilated elastin haploinsufficient newborn mice.
American journal of physiology. Lung cellular and molecular physiology (2014-12-30)
Anne Hilgendorff, Kakoli Parai, Robert Ertsey, Edwin Navarro, Noopur Jain, Francis Carandang, Joanna Peterson, Lucia Mokres, Carlos Milla, Stefanie Preuss, Miguel Alejandre Alcazar, Suleman Khan, Juliet Masumi, Nancy Ferreira-Tojais, Sana Mujahid, Barry Starcher, Marlene Rabinovitch, Richard Bland
ZUSAMMENFASSUNG
Elastin plays a pivotal role in lung development. We therefore queried if elastin haploinsufficient newborn mice (Eln(+/-)) would exhibit abnormal lung structure and function related to modified extracellular matrix (ECM) composition. Because mechanical ventilation (MV) has been linked to dysregulated elastic fiber formation in the newborn lung, we also asked if elastin haploinsufficiency would accentuate lung growth arrest seen after prolonged MV of neonatal mice. We studied 5-day-old wild-type (Eln(+/+)) and Eln(+/-) littermates at baseline and after MV with air for 8-24 h. Lungs of unventilated Eln(+/-) mice contained ∼50% less elastin and ∼100% more collagen-1 and lysyl oxidase compared with Eln(+/+) pups. Eln(+/-) lungs contained fewer capillaries than Eln(+/+) lungs, without discernible differences in alveolar structure. In response to MV, lung tropoelastin and elastase activity increased in Eln(+/+) neonates, whereas tropoelastin decreased and elastase activity was unchanged in Eln(+/-) mice. Fibrillin-1 protein increased in lungs of both groups during MV, more in Eln(+/-) than in Eln(+/+) pups. In both groups, MV caused capillary loss, with larger and fewer alveoli compared with unventilated controls. Respiratory system elastance, which was less in unventilated Eln(+/-) compared with Eln(+/+) mice, was similar in both groups after MV. These results suggest that elastin haploinsufficiency adversely impacts pulmonary angiogenesis and that MV dysregulates elastic fiber integrity, with further loss of lung capillaries, lung growth arrest, and impaired respiratory function in both Eln(+/+) and Eln(+/-) mice. Paucity of lung capillaries in Eln(+/-) newborns might help explain subsequent development of pulmonary hypertension previously reported in adult Eln(+/-) mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
Xylazin, ≥99%
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Serin, European Pharmacopoeia (EP) Reference Standard