Merck
Trabeculotomy ab interno with internal limiting membrane forceps for open-angle glaucoma.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (2014-03-29)
Houmei Nakasato, Riyo Uemoto, Masaru Isozaki, Akira Meguro, Tatsukata Kawagoe, Nobuhisa Mizuki
ZUSAMMENFASSUNG
To describe a new technique to perform trabeculotomy ab interno on eyes with open-angle glaucoma (OAG). This was a retrospective study. We inserted a 25-gauge forceps that is usually used for internal limiting membrane peeling into the anterior chamber, and grasped and pulled the inner wall of Schlemm's canal away from the canal. The inner wall of Schlemm's canal was stripped for about 100° to 120° in 26 eyes of 23 patients. The intraocular pressure (IOP) and number of glaucoma medications were recorded before, and 1 day, 1 week, 2 weeks, and 1, 3, 6, 10, 12, 15, 17, 19, 24, 27, 30, and 33 months after the surgery. The intra- and postoperative complications were recorded. The mean ± standard deviation of the preoperative IOP was 20.0 ± 6.8 mmHg with a range from 10 to 38 mmHg (n = 26). The IOP was significantly reduced (P < 0.05; paired t-tests) at 1 week, 2 weeks, and 1, 3, 6, 10, 12, 15, 17, 24, 19, 27, 30, and 33 months after the surgery. The mean preoperative number of glaucoma medications was significantly reduced (P < 0.001; paired t-tests) at 1 week, 2 weeks, and 1, 3, 6, 10, 12, 15, 17, 19, 24, 27, 30, and 33 months after the surgery. No vision-threatening complications were found in any of the cases, but there were blood clots in the anterior chamber postoperatively in 92.3 % of the cases. Trabeculotomy ab interno for OAG is effective but with some minor complications. A larger number of patients with longer follow-up periods are needed to determine the long-term effectiveness of this procedure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Pilocarpin -hydrochlorid, ≥99% (titration), powder
Supelco
Betamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Betamethason, ≥98%
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
USP
Betamethason, United States Pharmacopeia (USP) Reference Standard
Betamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Betamethason, VETRANAL®, analytical standard
Pilocarpin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard