Merck
  • Calorimetric investigation of diclofenac drug binding to a panel of moderately glycated serum albumins.
Calorimetric investigation of diclofenac drug binding to a panel of moderately glycated serum albumins.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-04-23)
Venkata S K Indurthi, Estelle Leclerc, Stefan W Vetter
ZUSAMMENFASSUNG
Glycation alters the drug binding properties of serum proteins and could affect free drug concentrations in diabetic patients with elevated glycation levels. We investigated the effect of bovine serum albumin glycation by eight physiologically relevant glycation reagents (glucose, ribose, carboxymethyllysine, acetoin, methylglyoxal, glyceraldehyde, diacetyl and glycolaldehyde) on diclofenac drug binding. We used this non-steroidal anti-inflammatory drug diclofenac as a paradigm for acidic drugs with high serum binding and because of its potential cardiovascular risks in diabetic patients. Isothermal titration calorimetry showed that glycation reduced the binding affinity Ka of serum albumin and diclofenac 2 to 6-fold by reducing structural rigidity of albumin. Glycation affected the number of drug binding sites in a glycation reagent dependent manner and lead to a 25% decrease for most reagent, expect for ribose, with decreased by 60% and for the CML-modification, increased the number of binding sites by 60%. Using isothermal titration calorimetry and differential scanning calorimetry we derived the complete thermodynamic characterization of diclofenac binding to all glycated BSA samples. Our results suggest that glycation in diabetic patients could significantly alter the pharmacokinetics of the widely used over-the-counter NSDAI drug diclofenac and with possibly negative implications for patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Glyoxylsäure -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard