Merck
  • Development of an optode for detection of trace amounts of Hg2+ in different real samples based on immobilization of novel tetradentate Schiff bases bearing two thiol groups in PVC membrane.
Development of an optode for detection of trace amounts of Hg2+ in different real samples based on immobilization of novel tetradentate Schiff bases bearing two thiol groups in PVC membrane.
Journal of fluorescence (2014-02-13)
Ayman A Abdel Aziz, Soha F Mohammed, Magdy M El Gamel
ZUSAMMENFASSUNG
A very sensitive and reversible optical chemical sensor based on a novel tetradentate Schiff base namely N.N(/)bis(2-aminothiophenol)benzene-1,2-dicarboxaldehyde (ATBD) immobilized within a plasticized PVC film for Hg(2+) determination is described. At optimum conditions (i.e. pH 6.0), the proposed sensor displayed a linear response to Hg(2+) over 1.0 × 10(-10) - 1.0 × 10(-2) mol L(-1) with a limit of detection of 7.23 × 10(-11) mol L(-1) (0.0145 μgL(-1)). Moreover, the results revealed that, under batch condition, the sensor is fully reversible within a response time ~ 35 s. In addition to its high stability and reproducibility, the sensor showed good selectivity towards Hg(2+) ion with respect to common metal cations. The sensor was successfully applied for determination of Hg(2+) ion in some real samples, including hair, urine and well water samples. The results were in good correlation with the data obtained using cold vapor atomic absorption spectrometry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Phthaldialdehyd, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phthaldialdehyd, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Phthaldialdehyd, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Supelco
Bis(2-ethylhexyl)adipat, 99%
Sigma-Aldrich
2-Aminothiophenol, 99%
Sigma-Aldrich
2-Nitrophenyl-octylether, 99%
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Nitrophenyl-octylether, Selectophore, ≥99.0%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Bis(2-ethylhexyl)adipat, ≥97.0% (GC)