Merck
  • A three-dimensional multiporous fibrous scaffold fabricated with regenerated spider silk protein/poly(l-lactic acid) for tissue engineering.
A three-dimensional multiporous fibrous scaffold fabricated with regenerated spider silk protein/poly(l-lactic acid) for tissue engineering.
Journal of biomedical materials research. Part A (2014-05-16)
Qiaozhen Yu, Chengjun Sun
ZUSAMMENFASSUNG
An axially aligned three-dimensional (3-D) fibrous scaffold was fabricated with regenerated spider silk protein (RSSP)/poly (l-lactic acid) (PLLA) through electrospinning and post treatment. The morphology, mechanical and degradation properties of the scaffold were controlled through the weight ratio of RSSP to PLLA, the thickness of the scaffold and the treatment time. The scaffold with a weight ratio of 2:3 (RSSP:PLLA) had a nanoleaves-on-nanofibers hierarchical nanostructure; the length and thickness of the nanoleaves were about 400 and 30 nm, respectively. The holes of the scaffolds ranged from hundreds of nanometers to several microns. The scaffold showed an ideal mechanical property that it was stiff when dry, but became soft once hydrated in the culture medium. Its degradation rate was very slow in the first 2 months, and then accelerated in the following 2 months. The pH values of the degradation mediums of all the samples remained in the range of 7.40-7.12 during degradation for 6 months. It had good biocompatibility with PC 12 cells. The aligned hierarchical nanostructure could guide the directions of the axon extension. This scaffold has a potential application in Tissue Engineering and controlled release. This study provides a method to produce synthetic or natural biodegradable polymer scaffold with tailored morphology, mechanical, and degradation properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard