Merck
  • Inhibition of inflammatory response in LPS-induced macrophages by 9-KOTE and 13-KOTE produced by biotransformation.
Inhibition of inflammatory response in LPS-induced macrophages by 9-KOTE and 13-KOTE produced by biotransformation.
Enzyme and microbial technology (2014-04-16)
Tânia Petta, Adriana Secatto, Lúcia Helena Faccioli, Luiz Alberto Beraldo Moraes
ZUSAMMENFASSUNG
Lipid mediators such as the leukotrienes, resolvins and protectins have been considered excellent models for the development of new anti-inflammatory drugs, due to their high potentiality. Nevertheless, only tiny amounts are available from natural sources and they have to be prepared by total synthesis. It is known that besides chemical reagents, microorganisms can also promote fatty acid oxygenation, via enzymatic reactions. In this context, the aim of this work was to produce oxylipids analogues in structure to lipid mediators employing microbial biotransformation. To this end, α-linolenic acid (ALA) was biotransformed by the fungi Aspergillus niger into oxylipids with different levels of oxygenation within 24h or 48h. The anti-inflammatory potential of products were evaluated by means of NO and TNF-α quantification in LPS-stimulated RAW264.7 macrophage cell line which guided the isolation of the regioisomers at m/z [M-H](-) 291, 9-keto-10E,12Z,15Z-octadecatrienoic acid (9-KOTE) and 13-keto-9Z,11E,15Z-octadecatrienoic acid (13-KOTE). We showed that biotransformation represents a powerful strategy for the production of potentially interesting candidates for development of anti-inflammation therapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, suitable for microscopy
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.