Merck
  • Chemical fingerprint analysis for quality control and identification of Ziyang green tea by HPLC.
Chemical fingerprint analysis for quality control and identification of Ziyang green tea by HPLC.
Food chemistry (2014-10-14)
Xiaoye He, Jianke Li, Wei Zhao, Run Liu, Lin Zhang, Xianghong Kong
ZUSAMMENFASSUNG
A simple and reliable HPLC fingerprint method was developed and validated for the quality control and identification of Ziyang green tea. Ten batches of Ziyang green tea collected from different plantations in Shaanxi Ziyang of China were used to establish the fingerprint. The feasibility and advantages of the used HPLC fingerprint were verified for its similarity evaluation by systematically comparing chromatograms with professional analytical software recommended by State Food and Drug Administration (SFDA) of China. The similarities of the fingerprints of 10 batches of tea samples were all more than 0.981. Additionally, simultaneous quantification of 10 major bioactive ingredients in the tea samples was conducted to interpret the consistency of the quality test. The results indicated that the HPLC fingerprint as a characteristic distinguishing method combining similarity evaluation and quantification analysis can be successfully used to assess the quality and to identify the authenticity of Ziyang green tea.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.