Merck
  • Functionally reciprocal mutations of the prolactin signalling pathway define hairy and slick cattle.
Functionally reciprocal mutations of the prolactin signalling pathway define hairy and slick cattle.
Nature communications (2014-12-19)
Mathew D Littlejohn, Kristen M Henty, Kathryn Tiplady, Thomas Johnson, Chad Harland, Thomas Lopdell, Richard G Sherlock, Wanbo Li, Steven D Lukefahr, Bruce C Shanks, Dorian J Garrick, Russell G Snell, Richard J Spelman, Stephen R Davis
ZUSAMMENFASSUNG
Lactation, hair development and homeothermy are characteristic evolutionary features that define mammals from other vertebrate species. Here we describe the discovery of two autosomal dominant mutations with antagonistic, pleiotropic effects on all three of these biological processes, mediated through the prolactin signalling pathway. Most conspicuously, mutations in prolactin (PRL) and its receptor (PRLR) have an impact on thermoregulation and hair morphology phenotypes, giving prominence to this pathway outside of its classical roles in lactation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinon, ≥99.0% (GC)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinon, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinon, reagent grade
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis