Merck
Urinary ADAM12 and MMP-9/NGAL complex detect the presence of gastric cancer.
Cancer prevention research (Philadelphia, Pa.) (2015-01-17)
Takaya Shimura, Adelle Dagher, Monisha Sachdev, Masahide Ebi, Tamaki Yamada, Tomonori Yamada, Takashi Joh, Marsha A Moses
ZUSAMMENFASSUNG
Although the early diagnosis of gastric cancer provides the opportunity for curative endoscopic resection, comprehensive screening endoscopy would be invasive and expensive. To date, there is a complete absence of clinically useful gastric cancer biomarkers. With the goal of discovering noninvasive biomarkers for the early diagnosis of gastric cancer, we have conducted a case-control study using urine samples from individuals with gastric cancer versus healthy control samples. Of the enrolled 106 patients from September, 2012 to April, 2013, a cohort of 70 patients composed of 35 patients with gastric cancer and 35 age- and sex-matched healthy controls was analyzed. The gastric cancer group was composed of stage IA of 62.9% (22/35). The urinary levels of MMP-9/NGAL complex (uMMP-9/NGAL) and ADAM12 (uADAM12) were significantly higher in the gastric cancer group compared with the healthy control group as determined by monospecific ELISAs (uMMP-9/NGAL: median, 85 pg/mL vs. 0 pg/mL; P = 0.020; uADAM12: median, 3.35 ng/mL vs. 1.44 ng/mL; P < 0.001). Multivariate analysis demonstrated that both uMMP-9/NGAL and uADAM12 were significant, independent diagnostic biomarkers for gastric cancer. Moreover, MMP-9/NGAL activity was significantly elevated as determined by gelatin zymography. The combination of uMMP-9/NGAL with uADAM12 distinguished between control samples and gastric cancer samples with an AUC of 0.825 (P < 0.001) in an ROC analysis. Significantly, immunohistochemical analyses demonstrated a high coexpression of MMP-9 and NGAL (P < 0.001) and high expression of ADAM12 (P < 0.001) in gastric cancer tissues compared with adjacent normal tissues (N = 35). In summary, uMMP-9/NGAL and uADAM12 are potential noninvasive biomarkers for gastric cancer, including early-stage disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.