Merck
  • In Vivo skin hydration and anti-erythema effects of Aloe vera, Aloe ferox and Aloe marlothii gel materials after single and multiple applications.
In Vivo skin hydration and anti-erythema effects of Aloe vera, Aloe ferox and Aloe marlothii gel materials after single and multiple applications.
Pharmacognosy magazine (2014-07-06)
Lizelle T Fox, Jeanetta du Plessis, Minja Gerber, Sterna van Zyl, Banie Boneschans, Josias H Hamman
ZUSAMMENFASSUNG
To investigate the skin hydrating and anti-erythema activity of gel materials from Aloe marlothii A. Berger and A. ferox Mill. in comparison to that of Aloe barbadensis Miller (Aloe vera) in healthy human volunteers. Aqueous solutions of the polisaccharidic fractions of the selected aloe leaf gel materials were applied to the volar forearm skin of female subjects. The hydration effect of the aloe gel materials were measured with a Corneometer(®) CM 825, Visioscan(®) VC 98 and Cutometer(®) dual MPA 580 after single and multiple applications. The Mexameter(®) MX 18 was used to determine the anti-erythema effects of the aloe material solutions on irritated skin areas. The A. vera and A. marlothii gel materials hydrated the skin after a single application, whereas the A. ferox gel material showed dehydration effects compared to the placebo. After multiple applications all the aloe materials exhibited dehydration effects on the skin. Mexameter(®) readings showed that A. vera and A. ferox have anti-erythema activity similar to that of the positive control group (i.e. hydrocortisone gel) after 6 days of treatment. The polysaccharide component of the gel materials from selected aloe species has a dehydrating effect on the skin after multiple applications. Both A. vera and A. ferox gel materials showed potential to reduce erythema on the skin similar to that of hydrocortisone gel.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Deuteriumoxid, filtered, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Deuteriumoxid, 70 atom % D
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Deuteriumoxid, 60 atom % D
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D