Merck
  • Lineage-related cytotoxicity and clonogenic profile of 1,4-benzoquinone-exposed hematopoietic stem and progenitor cells.
Lineage-related cytotoxicity and clonogenic profile of 1,4-benzoquinone-exposed hematopoietic stem and progenitor cells.
Toxicology and applied pharmacology (2015-02-04)
Paik Wah Chow, Zariyantey Abdul Hamid, Kok Meng Chan, Salmaan Hussain Inayat-Hussain, Nor Fadilah Rajab
ZUSAMMENFASSUNG
Hematopoietic stem cells (HSCs) and hematopoietic progenitor cells (HPCs) are sensitive targets for benzene-induced hematotoxicity and leukemogenesis. The impact of benzene exposure on the complex microenvironment of HSCs and HPCs remains elusive. This study aims to investigate the mechanism linking benzene exposure to targeting HSCs and HPCs using phenotypic and clonogenic analyses. Mouse bone marrow (BM) cells were exposed ex vivo to the benzene metabolite, 1,4-benzoquinone (1,4-BQ), for 24h. Expression of cellular surface antigens for HSC (Sca-1), myeloid (Gr-1, CD11b), and lymphoid (CD45, CD3e) populations were confirmed by flow cytometry. The clonogenicity of cells was studied using the colony-forming unit (CFU) assay for multilineage (CFU-GM and CFU-GEMM) and single-lineage (CFU-E, BFU-E, CFU-G, and CFU-M) progenitors. 1,4-BQ demonstrated concentration-dependent cytotoxicity in mouse BM cells. The percentage of apoptotic cells increased (p < 0.05) following 1,4-BQ exposure. Exposure to 1,4-BQ showed no significant effect on CD3e(+) cells but reduced the total counts of Sca-1(+), CD11b(+), Gr-1(+), and CD45(+) cells at 7 and 12 μM (p < 0.05). Furthermore, the CFU assay showed reduced (p < 0.05) clonogenicity in 1,4-BQ-treated cells. 1,4-BQ induced CFU-dependent cytotoxicity by significantly inhibiting colony growth for CFU-E, BFU-E, CFU-G, and CFU-M starting at a low concentration of exposure (5μM); whereas for the CFU-GM and CFU-GEMM, the inhibition of colony growth was remarkable only at 7 and 12μM of 1,4-BQ, respectively. Taken together, 1,4-BQ caused lineage-related cytotoxicity in mouse HPCs, demonstrating greater toxicity in single-lineage progenitors than in those of multi-lineage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Supelco
Benzol, analytical standard
Sigma-Aldrich
Benzol, anhydrous, 99.8%
USP
1,4-Benzochinon, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
p-Benzochinon, reagent grade, ≥98%
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Propidiumjodid -Lösung
Supelco
1,4-Benzochinon, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure, technical grade
Supelco
p-Benzochinon, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure