Merck
Surface modification-induced phase transformation of hexagonal close-packed gold square sheets.
Nature communications (2015-03-15)
Zhanxi Fan, Xiao Huang, Yu Han, Michel Bosman, Qingxiao Wang, Yihan Zhu, Qing Liu, Bing Li, Zhiyuan Zeng, Jumiati Wu, Wenxiong Shi, Shuzhou Li, Chee Lip Gan, Hua Zhang
ZUSAMMENFASSUNG
Conventionally, the phase transformation of inorganic nanocrystals is realized under extreme conditions (for example, high temperature or high pressure). Here we report the complete phase transformation of Au square sheets (AuSSs) from hexagonal close-packed (hcp) to face-centered cubic (fcc) structures at ambient conditions via surface ligand exchange, resulting in the formation of (100)f-oriented fcc AuSSs. Importantly, the phase transformation can also be realized through the coating of a thin metal film (for example, Ag) on hcp AuSSs. Depending on the surfactants used during the metal coating process, two transformation pathways are observed, leading to the formation of (100)f-oriented fcc Au@Ag core-shell square sheets and (110)h/(101)f-oriented hcp/fcc mixed Au@Ag nanosheets. Furthermore, monochromated electron energy loss spectroscopy reveals the strong surface plasmon resonance absorption of fcc AuSS and Au@Ag square sheet in the infrared region. Our findings may offer a new route for the crystal-phase and shape-controlled synthesis of inorganic nanocrystals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Oleylamin, technical grade, 70%
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumborhydrid, ReagentPlus®, 99%
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline