Merck
  • Synthesis and characterization of the anticancer and metal binding properties of novel pyrimidinylhydrazone derivatives.
Synthesis and characterization of the anticancer and metal binding properties of novel pyrimidinylhydrazone derivatives.
Journal of inorganic biochemistry (2015-01-16)
Veronika F S Pape, Dóra Türk, Pál Szabó, Michael Wiese, Eva A Enyedy, Gergely Szakács
ZUSAMMENFASSUNG
Three novel pyrimidinylhydrazones substituted at either the aromatic moiety or at the imine carbon atom were synthesized and characterized by standard analytical methods. All compounds were found to be toxic in the micro- to submicromolar range against a diverse panel of cancer cell lines including multidrug resistant (MDR) derivatives expressing P-glycoprotein (Pgp). UV-visible spectrophotometry experiments demonstrated that the most active compound (3) forms highly stable complexes with iron(III) and copper(II) in a wide pH range with a stronger preference towards iron(III). The redox activity of the iron and copper complexes of ligand 3 was investigated using cyclic voltammetry and was tested with cellular reductants. The impact of reactive oxygen species (ROS) on the mechanism of toxicity was assessed using the ROS-sensitive cell permeable dye 2',7'-dichlorofluorescin diacetate (DCFDA). Our results demonstrate that the studied pyrimidinylhydrazones form redox-active iron and copper complexes that are capable of producing intracellular ROS, which might lead to cellular damage and cell death in cancer cells regardless of their resistance status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
2′,7′-Dichlorfluorescein-Diacetat, ≥97%
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Tetrabutylammoniumchlorid, ≥97.0% (NT)
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
SAFC
L-Glutamin
Sigma-Aldrich
Ferrocen, 98%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis