Merck
  • Protein kinase C promotes cardiac fibrosis and heart failure by modulating galectin-3 expression.
Protein kinase C promotes cardiac fibrosis and heart failure by modulating galectin-3 expression.
Biochimica et biophysica acta (2014-12-10)
Xiang Song, Xiaoqian Qian, Ming Shen, Rong Jiang, Mary B Wagner, Guoliang Ding, Guangping Chen, Baozhong Shen
ZUSAMMENFASSUNG
Protein kinase C (PKC) and galectin-3 are two important mediators that play a key pathogenic role in cardiac hypertrophy and heart failure (HF). However, the molecular mechanisms and signaling pathways are not fully understood. In this study, we explored the relationship between and roles of PKC-α and galectin-3 in the development of HF. We found that activation of PKC by phorbol dibutyrate (PDB) increased galectin-3 expression by ~180%, as well as collagen I and fibronection accumulation in cultured HL-1 cardiomyocytes. Over-expression of galectin-3 in HL-1 cells increased collagen I protein production. Inhibition of galectin-3 by β-lactose blocked PDB-induced galectin-3 and collagen production, indicating that galectin-3 mediates PKC-induced cardiac fibrosis. In rats subjected to pulmonary artery banding (PAB) to induce right ventricular HF, galectin-3 was increased by ~140% in the right ventricle and also by ~240% in left ventricle compared to control. The elevated galectin-3 is consistent with an increase of total and activated (phosphorylated) PKC-α, α-SMA and collagen I. Finally, we extended our findings to examine the role of angiotensin II (Ang II), which activates the PKC pathway and contributes to cardiac fibrosis and the development of HF. We found that Ang II activated the PKC-α pathway and increased galectin-3 expression and collagen production. This study provides a new insight into the molecular mechanisms of HF mediated by PKC-α and galectin-3. PKC-α promotes cardiac fibrosis and HF by stimulation of galectin-3 expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
β-Laktose, ≤30% α-anomer basis, ≥99% total lactose basis
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Phorbol-12,13-dibutyrat, ≥98% (TLC), powder
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chelerythrinchlorid, ≥95% (TLC), powder
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland