Merck
  • Solvothermal-assisted liquid-phase exfoliation of graphite in a mixed solvent of toluene and oleylamine.
Solvothermal-assisted liquid-phase exfoliation of graphite in a mixed solvent of toluene and oleylamine.
Nanoscale research letters (2015-06-10)
Dinh Khoi Dang, Eui Jung Kim
ZUSAMMENFASSUNG
We report an effective method for producing graphene sheets using solvothermal-assisted exfoliation of graphite in a mixed solvent of toluene and oleylamine. The mixed solvent of toluene and oleylamine produces higher yield of graphene than its constituents, oleylamine and toluene. The oleylamine molecules with its long chain enwrap the graphene sheets efficiently, while toluene helps the oleylamine molecules become more flexible and easily intercalate into the edge of graphite. The prepared graphene sheets have a high quality, and the concentration of graphene in the dispersion is as high as 0.128 mg mL(-1). The high-quality graphene sheets obtained in this work make them suitable for application in many fields such as energy-storage materials and polymer composites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Kaliumbromid, ≥99% trace metals basis
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ReagentPlus®, 99%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Oleylamin, technical grade, 70%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, anhydrous, 99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, suitable for HPLC, ≥99%
Sigma-Aldrich
Kaliumbromid, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, biotech. grade, ≥99.7%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumbromid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
SAFC
Oleylamin, ≥98% (primary amine)
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumbromid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumbromid, BioUltra, ≥99.5% (AT)
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumbromid, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
1-Methyl-2-pyrrolidon, analytical standard
Sigma-Aldrich
Kaliumbromid, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Kaliumbromid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumbromid, BioXtra, ≥99.0%