Merck
  • Gene silencing of Rac1 with RNA interference mediated by ultrasound and microbubbles in human LoVo cells: evaluation of cell invasion inhibition and metastatic.
Gene silencing of Rac1 with RNA interference mediated by ultrasound and microbubbles in human LoVo cells: evaluation of cell invasion inhibition and metastatic.
Journal of drug targeting (2015-02-13)
Fang Nie, Xiao-Feng Wang, Shi-Yi Zhao, Lan Bu, Xue-Hui Liu
ZUSAMMENFASSUNG
The objective of this study was to assess the change of cytoskeleton and cell cycle in LoVo (human colorectal cancer) cell via gene silencing of Rac1 with RNA interference mediated by microbubble (SonoVue) and ultrasound (US). The compound of plasmid Rac1-shRNA, LoVo cells, and SonoVue was exposed to US (1 MHz, 2 W/cm(2), 5 min). The expression of Rac1 mRNA and Rac1 protein was detected by RT-PCR and Western blot. Cytoskeleton was taken by confocal microscope in a random fashion. Cell invasion was assayed using modified Boyden chambers, and cell cycle and apoptosis were analyzed by flow cytometry in LoVo cells. Rac1 gene is overexpressed in human colorectal cancer cells, gene silencing of Rac1 with RNA interference mediated by microbubble and US strongly inhibited lamellipodia formation, cell invasion, and delayed cell cycle, as well as enhanced cell apoptosis of LoVo cells in vitro. Silencing Rac1 gene mediated by microbubble and US may become a new treatment option for the inhibition of the invasion and metastasis of colorectal cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Propidiumjodid -Lösung
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.