Merck
  • KCTD10 is involved in the cardiovascular system and Notch signaling during early embryonic development.
KCTD10 is involved in the cardiovascular system and Notch signaling during early embryonic development.
PloS one (2014-11-18)
Kaiqun Ren, Jing Yuan, Manjun Yang, Xiang Gao, Xiaofeng Ding, Jianlin Zhou, Xingwang Hu, Jianguo Cao, Xiyun Deng, Shuanglin Xiang, Jian Zhang
ZUSAMMENFASSUNG
As a member of the polymerase delta-interacting protein 1 (PDIP1) gene family, potassium channel tetramerisation domain-containing 10 (KCTD10) interacts with proliferating cell nuclear antigen (PCNA) and polymerase δ, participates in DNA repair, DNA replication and cell-cycle control. In order to further investigate the physiological functions of KCTD10, we generated the KCTD10 knockout mice. The heterozygous KCTD10(+/-) mice were viable and fertile, while the homozygous KCTD10(-/-) mice showed delayed growth from E9.0, and died at approximately E10.5, which displayed severe defects in angiogenesis and heart development. Further study showed that VEGF induced the expression of KCTD10 in a time- and dose-dependent manner. Quantitative real-time PCR and western blotting results revealed that several key members in Notch signaling were up-regulated either in KCTD10-deficient embryos or in KCTD10-silenced HUVECs. Meanwhile, the endogenous immunoprecipitation (IP) analysis showed that KCTD10 interacted with Cullin3 and Notch1 simultaneously, by which mediating Notch1 proteolytic degradation. Our studies suggest that KCTD10 plays crucial roles in embryonic angiogenesis and heart development in mammalians by negatively regulating the Notch signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Kctd10
Sigma-Aldrich
MISSION® esiRNA, targeting human KCTD10