Merck
  • Sulfonated polystyrene magnetic nanobeads coupled with immunochromatographic strip for clenbuterol determination in pork muscle.
Sulfonated polystyrene magnetic nanobeads coupled with immunochromatographic strip for clenbuterol determination in pork muscle.
Talanta (2014-08-17)
Kesheng Wu, Liang Guo, Wei Xu, Hengyi Xu, Zoraida P Aguilar, Guomao Xu, Weihua Lai, Yonghua Xiong, Yiqun Wan
ZUSAMMENFASSUNG
A magnetic solid-phase extraction method (MSPE) was developed to pre-concentrate and cleanup clenbuterol (CLE) from pork muscle. Novel sulfonated polystyrene magnetic nanobeads (spMNBs) were synthesized via a one-pot emulsion copolymerization method by using divinylbenzene, styrene, and sodium styrene sulfonate in the presence of oleic acid-modified and 10-undecylenic acid-modified magnetic ferrofluid. The resulting spMNBs exhibited high adsorption efficiency for CLE and for 10 other common beta-adrenergic agonists, namely, brombuterol, ractopamine, tulobuterol, bambuterol, cimbuterol, mabuterol, clorprenaline, penbutolol, salbutamol, and cimaterol. The adsorption behavior of the spMNBs for CLE was described by the Langmuir equation with a maximum adsorption capacity of 0.41 mg/g. Under the optimized parameters, the extraction of CLE from 0.5 g of pork muscle required 25mg of the spMNBs at a shortened adsorption time (0.5 min). The proposed MSPE was coupled with colloidal gold nanoparticle-based immunochromatographic assay (MSPE-AuNPIA) for the quantitative detection of CLE residue in pork muscle. The limit of detection and limit of quantification for the pork muscle were 0.10 and 0.24 ng/g, respectively. The intra-day and inter-day assay recoveries at three CLE spiked concentrations ranged from 92.5% to 98.1%, with relative standard deviations ranging from 3.2% to 13.0%. The results of MSPE-AuNPIA were confirmed by liquid chromatography-tandem mass spectrometry (LC-MS/MS). The CLE values obtained with MSPE-AuNPIA agreed with those obtained with LC-MS/MS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumpersulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Kaliumperoxodisulfat, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Styrol, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Divinylbenzol, technical grade, 55%
Sigma-Aldrich
Divinylbenzol, technical grade, 80%
Sigma-Aldrich
Kaliumpersulfat, 99.99% trace metals basis
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Natrium-4-vinylbenzolsulfonat, technical, ≥90% (T)
Supelco
Styrol, analytical standard
Supelco
Oleinsäure, analytical standard
Sigma-Aldrich
Salbutamol
Supelco
Salbutamol, VETRANAL®, analytical standard
Supelco
Cimaterol, VETRANAL®, analytical standard
Salbutamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleinsäure, Selectophore, ≥99%
Oleinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Cimbuterol, VETRANAL®, analytical standard