Merck
  • Smooth muscle specific disruption of the endothelin-A receptor in mice reduces arterial pressure, and vascular reactivity and affects vascular development.
Smooth muscle specific disruption of the endothelin-A receptor in mice reduces arterial pressure, and vascular reactivity and affects vascular development.
Life sciences (2014-01-15)
Anthony J Donato, Lisa A Lesniewski, Deborah Stuart, Ashley E Walker, Grant Henson, Lise Sorensen, Dean Li, Donald E Kohan
ZUSAMMENFASSUNG
The role of vascular smooth muscle endothelin A receptors (ETA) in development and normal physiology remains incompletely understood. To address this, mice were generated with smooth muscle-specific knockout (KO) of ETA. Mice were homozygous for loxP-flanked exons 6-8 of the EDNRA gene (floxed) or were also hemizygous for a transgene expressing Cre recombinase under control of the smooth muscle-specific SM22 promoter (KO mice). Genotyping at 17 days postnatal yielded a 10:1 ratio of floxed:KO mice. Smooth muscle actin staining of embryos at day E10.5 revealed increased tortuosity in dorsal aortae while E12.5 embryos had mandibular, vascular and thymic abnormalities. Mice surviving to weaning developed and bred normally. ETA KO mice aged 2-3 months manifested EDNRA gene recombination in all organs tested. Aortas from KO mice had a >90% reduction in ETA mRNA content, but no differences in ET-1 or ETB mRNA levels. Addition of 0.01-100 nM ET-1 to isolated femoral arteries from floxed, but not KO, mice dose-dependently decreased vessel diameter (up to 80% reduction in the presence of ETB blockade). Intravenous infusion of ET-1 into floxed, but not KO, mice increased mean arterial pressure (MAP) (by ~10 mm Hg). Telemetric analysis revealed decreased MAP in KO mice (reduced by ~7-10 mm Hg) when fed a high salt diet. Smooth muscle ETA is important for normal vascular, mandibular and thymic development and is involved in the maintenance of arterial pressure under physiological conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Benzylalkohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzylalkohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Supelco
Benzylalkohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanolamin, ≥99%
Sigma-Aldrich
Nickel(II)-chlorid, 98%
USP
Benzylalkohol, United States Pharmacopeia (USP) Reference Standard
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Ethanolamin, ≥98%
Sigma-Aldrich
Ethanolamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Benzylalkohol, anhydrous, 99.8%
Sigma-Aldrich
Benzylalkohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Ethanolamin, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzylbenzoat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Nickel(II)-chlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Benzylalkohol, ≥99%, FCC, FG
Supelco
Benzylalkohol, analytical standard
Sigma-Aldrich
Benzylbenzoat, ≥99%, FCC, FG
Sigma-Aldrich
Ethanolamin, liquid, BioReagent, suitable for cell culture, ≥98%
USP
Benzylbenzoat, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzylbenzoat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzylalkohol, natural, ≥98%, FG
Benzylalkohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzylbenzoat, natural, ≥99%, FG