Merck
  • Oxidative damage to mitochondria at the nodes of Ranvier precedes axon degeneration in ex vivo transected axons.
Oxidative damage to mitochondria at the nodes of Ranvier precedes axon degeneration in ex vivo transected axons.
Experimental neurology (2014-06-29)
Helena Bros, Jason M Millward, Friedemann Paul, Raluca Niesner, Carmen Infante-Duarte
ZUSAMMENFASSUNG
Oxidative stress and mitochondrial dysfunction appear to contribute to axon degeneration in numerous neurological disorders. However, how these two processes interact to cause axonal damage-and how this damage is initiated-remains unclear. In this study we used transected motor axons from murine peripheral roots to investigate whether oxidative stress alters mitochondrial dynamics in myelinated axons. We show that the nodes of Ranvier are the initial sites of mitochondrial damage induced by oxidative stress. There, mitochondria became depolarized, followed by alterations of the external morphology and disruption of the cristae, along with reduced mitochondrial transport. These mitochondrial changes expanded from the nodes of Ranvier bidirectionally towards both internodes and eventually affected the entire mitochondrial population in the axon. Supplementing axonal bioenergetics by applying nicotinamide adenine dinucleotide and methyl pyruvate, rendered the mitochondria at the nodes of Ranvier resistant to these oxidative stress-induced changes. Importantly, this inhibition of mitochondrial damage protected the axons from degeneration. In conclusion, we present a novel ex vivo approach for monitoring mitochondrial dynamics within axons, which proved suitable for detecting mitochondrial changes upon exogenous application of oxidative stress. Our results indicate that the nodes of Ranvier are the site of initial mitochondrial damage in peripheral axons, and suggest that dysregulation of axonal bioenergetics plays a critical role in oxidative stress-triggered mitochondrial alterations and subsequent axonal injury. These novel insights into the mechanisms underlying axon degeneration may have implications for neurological disorders with a degenerative component.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Methylpyruvat, 90%, technical grade
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.