Merck
  • Nitrate (NO3(-)) and nitrite (NO2(-)) are endocrine disruptors to downregulate expression of tyrosine hydroxylase and motor behavior through conversion to nitric oxide in early development of zebrafish.
Nitrate (NO3(-)) and nitrite (NO2(-)) are endocrine disruptors to downregulate expression of tyrosine hydroxylase and motor behavior through conversion to nitric oxide in early development of zebrafish.
Biochemical and biophysical research communications (2014-09-01)
Meshkatul Jannat, Ratu Fatimah, Mitsuyo Kishida
ZUSAMMENFASSUNG
With a view to consider the increasing concern over nitrogen pollution in the aquatic environment, we investigated effects of nitrate (NO3(-)) and nitrite (NO2(-)) on the activity of dopaminergic neuron in zebrafish embryos and larvae. Both nitrate and nitrite exposure decreased the expression of tyrosine hydroxylase (TH) in dopaminergic neurons at 48hpf. Only nitrite decreased the response to tactile stimulation at 72hpf, whereas both nitrate and nitrite decreased the swimming activity at 6dpf. When the embryos were exposed to nitrate or nitrite together with an estrogen receptor blocker (ICI 182,780), the decreases in TH expression and motor behavior caused by nitrate or nitrite alone were reversed suggesting the effects of nitrate and nitrite were mediated through estrogen receptor (ER). The result of co-incubation with an oxidoreductase inhibitor, diphenyleneiodonium, indicated the conversion to nitric oxide (NO) is likely to be responsible for the effects of nitrate and nitrite, which was further supported by the increased staining for NO after exposure. The present study demonstrates that nitrate and nitrite are neurotoxicants acting as an endocrine disruptor possibly through conversion to NO to downregulate the activity of dopaminergic neuron in early development of zebrafish.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumnitrit, ACS reagent, ≥96.0%
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumnitrit, meets analytical specification of FCC, 97-100.5%
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143095, Kaliumnitrat, traceable to primary standards (LGC)