Merck
Phenolic profiling in the pulp and peel of nine plantain cultivars (Musa sp.).
Food chemistry (2014-08-26)
Claudine Valérie Passo Tsamo, Marie-France Herent, Kodjo Tomekpe, Thomas Happi Emaga, Joëlle Quetin-Leclercq, Hervé Rogez, Yvan Larondelle, Christelle Andre
ZUSAMMENFASSUNG
The present study investigated the phenolic profiles of the pulp and peel of nine plantain cultivars and compared them to those of two dessert bananas of commercial interest (Grand Nain and Gros Michel), alongside a newly created hybrid, resistant to black sigatoka disease (F568). Identification and quantification of phenolic compounds were performed by means of HPLC-ESI-HR-MS and HPLC-DAD. Hydroxycinnamic acids, particularly ferulic acid-hexoside with 4.4-85.1 μg/g of dry weight, dominated in the plantain pulp and showed a large diversity among cultivars. Flavonol glycosides were predominant in plantain peels, rutin (242.2-618.7 μg/g of dry weight) being the most abundant. A principal component analysis on the whole data revealed that the phenolic profiles of the hybrid, the dessert bananas and the pure plantains differed from each other. Plantain pulps and peels appeared as good sources of phenolics, which could be involved in the health benefits associated with their current applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Isorhamnetin, ≥95.0% (HPLC)
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Supelco
trans-Ferulasäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Kaempferol, analytical standard
Supelco
Isorhamnetin, analytical standard
Ferulasäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulasäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland