Merck
Ellipticine derivative induces potent cytostatic effect in acute myeloid leukaemia cells.
Investigational new drugs (2014-08-12)
E G Russell, E C O'Sullivan, C M Miller, J Stanicka, F O McCarthy, T G Cotter
ZUSAMMENFASSUNG
A panel of novel ellipticine isomers were designed and synthesised with the aim of evaluating their anti-cancer effects on selected leukaemia cell lines. A preliminary NCI 60-cell screen demonstrated that these compounds display promising anti-tumour activity across a number of different cell types. We have consequently examined the effect of these derivatives in detail on the Acute Myeloid Leukaemia (AML) cell line, MV4-11. Cell cycle analyses revealed that the compounds had a range of distinctive cell cycle effects. 7-Hydroxyisoellipticine showed the most promise with respect to cytostatic activity. We demonstrated that this compound inhibited proliferation of leukaemia cells by preventing cells from progressing from G2 phase into mitosis over a period of 24 h at a concentration of 5 μM. Our research suggests that this is mediated by an induction of reactive oxygen species (ROS), which in turn activates the DNA damage response pathway. As a result of the activation of p53, cyclin B1 is inhibited. The induction of this pathway leads to apoptosis which is seen at 48 h using the same dose of 7-hydroxyisoellipticine. This study provides for the first time detailed cellular information on the potential use of isoellipticines as chemotherapeutic agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, suitable for microscopy
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar