Merck
  • Thrombospondin-1 deficiency causes a shift from fibroproliferative to inflammatory kidney disease and delays onset of renal failure.
Thrombospondin-1 deficiency causes a shift from fibroproliferative to inflammatory kidney disease and delays onset of renal failure.
The American journal of pathology (2014-08-12)
Michael Zeisberg, Björn Tampe, Valerie LeBleu, Desiree Tampe, Elisabeth M Zeisberg, Raghu Kalluri
ZUSAMMENFASSUNG
Thrombospondin-1 (TSP1) is a multifunctional matricellular protein known to promote progression of chronic kidney disease. To gain insight into the underlying mechanisms through which TSP1 accelerates chronic kidney disease, we compared disease progression in Col4a3 knockout (KO) mice, which develop spontaneous kidney failure, with that of Col4a3;Tsp1 double-knockout (DKO) mice. Decline of excretory renal function was significantly delayed in the absence of TSP1. Although Col4a3;Tsp1 DKO mice did progress toward end-stage renal failure, their kidneys exhibited distinct histopathological lesions, compared with creatinine level-matched Col4a3 KO mice. Although kidneys of both Col4a3 KO and Col4a3;Tsp1 DKO mice exhibited a widened tubulointerstitium, predominant lesions in Col4a3 KO kidneys were collagen deposition and fibroblast accumulation, whereas in Col4a3;Tsp1 DKO kidney inflammation was predominant, with less collagen deposition. Altered disease progression correlated with impaired activation of transforming growth factor-β1 (TGF-β1) in vivo and in vitro in the absence of TSP1. In summary, our findings suggest that TSP1 contributes to progression of chronic kidney disease by catalyzing activation of latent TGF-β1, resulting in promotion of a fibroproliferative response over an inflammatory response. Furthermore, the findings suggest that fibroproliferative and inflammatory lesions are independent entities, both of which contribute to decline of renal function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Phosphatase-Substrat, 5 mg tablets
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, tablet
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Phosphatase-Substrat, powder
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, tablet
Sigma-Aldrich
Phosphatase-Substrat, 40 mg tablets
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, tablet
Sigma-Aldrich
Phosphatase-Substrat, 100 mg capsules
Sigma-Aldrich
Phosphatase-Substrat, 40 mg capsules
Sigma-Aldrich
Phosphatasesubstrat, Suitable for manufacturing of diagnostic kits and reagents