Merck
Characterization of freeze-dried gallic acid/xyloglucan.
Drug development and industrial pharmacy (2013-11-16)
Namon Hirun, Tanatchaporn Sangfai, Vimon Tantishaiyakul
ZUSAMMENFASSUNG
Tamarind seed xyloglucan (TSX) is generally used for drug delivery systems. Gallic acid (GA) possesses various pharmacological activities. It has a good solubility and bioavailability but short half-life. To prepare a sustained-release of GA to overcome its relatively short half-life. GA was blended with TSX and freeze-dried. The physicochemical properties of freeze-dried GA and freeze-dried GA/TSX were characterized, and the release profiles of GA from these freeze-dried samples were investigated. All freeze-dried samples were characterized by PXRD, spectroscopic and thermal analyses. The dissolution studies were performed according to the United States Pharmacopeia (USP) XXX. According to FTIR, FT-Raman and (13)C CP/MAS NMR, the spectra of freeze-dried GA were similar to that of the anhydrous form. Nevertheless, DRIFTS and DSC were able to differentiate these two forms. The crystallinity of GA in the freeze-dried GA/TSX was the same as that of the freeze-dried GA. DSC indicates that there were interactions between GA and TSX. It was of interest that a freeze-dried sample with low amount of GA, 0.2% GA/1% TSX was mostly in an amorphous form. Moreover, all freeze-dried GA/TSX preparations demonstrated a sustained-release of GA compared to GA alone. The freeze-dried 1% GA/1% TSX provided the best sustained-release of GA of up to 240 min. TSX could change a crystal form of a small molecule to a mostly amorphous form. It was of importance that the freeze-dried GA/TSX could effectively retard the release of GA. These samples may be able to overcome the limitation for the therapeutic use of GA due to its short biological half-life.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Saccharose, European Pharmacopoeia (EP) Reference Standard
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Galactose, European Pharmacopoeia (EP) Reference Standard