Merck
  • Immunomodulatory effects of clove (Syzygium aromaticum) constituents on macrophages: in vitro evaluations of aqueous and ethanolic components.
Immunomodulatory effects of clove (Syzygium aromaticum) constituents on macrophages: in vitro evaluations of aqueous and ethanolic components.
Journal of immunotoxicology (2014-05-31)
Shaghayegh Pishkhan Dibazar, Shirin Fateh, Saeed Daneshmandi
ZUSAMMENFASSUNG
The present work sought to investigate potential suppressive effects on mouse macrophages by in vitro treatment with clove (Syzygium aromaticum) ethanolic extracted essential oil (containing eugenol) or its water-soluble extract. Using doses (ranging from 0.001-1000 µg/ml) of each material freshly prepared in the laboratory, cell survival and production of nitric oxide (NO), tumor necrosis factor (TNF)-α, interleukin (IL)-6, and IL-12 by the treated cells (that in all cases also had received LPS stimulation) were measured. Results indicated that, except at doses ≥100 µg/ml, viability was unaffected in all groups. NO release by LPS-stimulated macrophages was generally significantly suppressed by either material; in contrast, low (i.e. 0.001-1 µg/ml) doses of either extract class appeared to enhance NO release by non-LPS (unstimulated)-treated macrophages. Among LPS-stimulated cells, TNFα release was also significantly affected by each extract; the ethanolic extract was suppressive at all doses tested, while the aqueous material was so up to 1 µg/ml and then became stimulatory. In contrast, nearly every dose of either extract appeared to stimulate IL-6 release from the LPS-treated cells. Effects on IL-12 production were overall inconsistent; in general, the ethanolic extract tended to be stimulatory of production by the LPS-treated cells. The data for the aqueous material showed no discernable pattern of effect. The results suggest that clove extracts do not have a distinct cytotoxic activity, but do impart potential anti- and pro-oxidant effects in cells, depending on their concentrations and on the activation state of the macrophages themselves at the time of exposure to the extracts. The impact of the extracts on macrophage cytokine release also displays a pattern of dose-relatedness.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
SAFC
L-Glutamin
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylendiamin -dihydrochlorid, 98%
Supelco
Sulfanilamid Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Supelco
Sulfanilamid, VETRANAL®, analytical standard
Sulfanilamid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland