Merck
  • A whole recombinant yeast-based therapeutic vaccine elicits HBV X, S and Core specific T cells in mice and activates human T cells recognizing epitopes linked to viral clearance.
A whole recombinant yeast-based therapeutic vaccine elicits HBV X, S and Core specific T cells in mice and activates human T cells recognizing epitopes linked to viral clearance.
PloS one (2014-07-23)
Thomas H King, Charles B Kemmler, Zhimin Guo, Derrick Mann, Yingnian Lu, Claire Coeshott, Adam J Gehring, Antonio Bertoletti, Zi Z Ho, William Delaney, Anuj Gaggar, G Mani Subramanian, John G McHutchison, Shikha Shrivastava, Yu-Jin L Lee, Shyamasundaran Kottilil, Donald Bellgrau, Timothy Rodell, David Apelian
ZUSAMMENFASSUNG
Chronic hepatitis B infection (CHB) is characterized by sub-optimal T cell responses to viral antigens. A therapeutic vaccine capable of restoring these immune responses could potentially improve HBsAg seroconversion rates in the setting of direct acting antiviral therapies. A yeast-based immunotherapy (Tarmogen) platform was used to make a vaccine candidate expressing hepatitis B virus (HBV) X, surface (S), and Core antigens (X-S-Core). Murine and human immunogenicity models were used to evaluate the type and magnitude of HBV-Ag specific T cell responses elicited by the vaccine. C57BL/6J, BALB/c, and HLA-A*0201 transgenic mice immunized with yeast expressing X-S-Core showed T cell responses to X, S and Core when evaluated by lymphocyte proliferation assay, ELISpot, intracellular cytokine staining (ICS), or tumor challenge assays. Both CD4+ and CD8+ T cell responses were observed. Human T cells transduced with HBc18-27 and HBs183-91 specific T cell receptors (TCRs) produced interferon gamma (IFNγ following incubation with X-S-Core-pulsed dendritic cells (DCs). Furthermore, stimulation of peripheral blood mononuclear cells (PBMCs) isolated from CHB patients or from HBV vaccine recipients with autologous DCs pulsed with X-S-Core or a related product (S-Core) resulted in pronounced expansions of HBV Ag-specific T cells possessing a cytolytic phenotype. These data indicate that X-S-Core-expressing yeast elicit functional adaptive immune responses and supports the ongoing evaluation of this therapeutic vaccine in patients with CHB to enhance the induction of HBV-specific T cell responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thymidin, ≥99.0% (HPLC)
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysin-hydrochlorid -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland