Merck
Simulation model for transcervical laryngeal injection providing real-time feedback.
The Annals of otology, rhinology, and laryngology (2014-06-26)
Tiffiny A Ainsworth, James B Kobler, Gregory J Loan, James A Burns
ZUSAMMENFASSUNG
This study aimed to develop and evaluate a model for teaching transcervical laryngeal injections. A 3-dimensional printer was used to create a laryngotracheal framework based on de-identified computed tomography images of a human larynx. The arytenoid cartilages and intrinsic laryngeal musculature were created in silicone from clay casts and thermoplastic molds. The thyroarytenoid (TA) muscle was created with electrically conductive silicone using metallic filaments embedded in silicone. Wires connected TA muscles to an electrical circuit incorporating a cell phone and speaker. A needle electrode completed the circuit when inserted in the TA during simulated injection, providing real-time feedback of successful needle placement by producing an audible sound. Face validation by the senior author confirmed appropriate tactile feedback and anatomical realism. Otolaryngologists pilot tested the model and completed presimulation and postsimulation questionnaires. The high-fidelity simulation model provided tactile and audio feedback during needle placement, simulating transcervical vocal fold injections. Otolaryngology residents demonstrated higher comfort levels with transcervical thyroarytenoid injection on postsimulation questionnaires. This is the first study to describe a simulator for developing transcervical vocal fold injection skills. The model provides real-time tactile and auditory feedback that aids in skill acquisition. Otolaryngologists reported increased confidence with transcervical injection after using the simulator.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), beads
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, ≥350 μm particle size