Merck
Renewable-juglone-based high-performance sodium-ion batteries.
Advanced materials (Deerfield Beach, Fla.) (2015-03-03)
Hua Wang, Pengfei Hu, Jie Yang, Guangming Gong, Lin Guo, Xiaodong Chen
ZUSAMMENFASSUNG
A renewable-biomolecule-based electrode is developed through a facile synchronous reduction and self-assembly process, without any binder or additional conductive agent. The hybridized electrodes can be fabricated with arbitrary size and shape and exhibit superior capacity and cycle performance. The renewable-biomaterial-based high-performance electrodes will hold a place in future energy-storage devices.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylcarbonat, anhydrous, ≥99%
Sigma-Aldrich
Ethylencarbonat, 98%
Sigma-Aldrich
Dimethylcarbonat, ReagentPlus®, 99%