Merck
Glucocorticoids induce autophagy in rat bone marrow mesenchymal stem cells.
Molecular medicine reports (2014-12-18)
Long Wang, Jing Fan, Yan-Shui Lin, Yun-Shan Guo, Bo Gao, Qi-Yue Shi, Bo-Yuan Wei, Li Chen, Liu Yang, Jian Liu, Zhuo-Jing Luo
ZUSAMMENFASSUNG
Glucocorticoid‑induced osteoporosis (GIOP) is a widespread clinical complication following glucocorticoid therapy. This irreversible damage to bone‑forming and ‑resorbing cells is essential in the pathogenesis of osteoporosis. Autophagy is a physiological process involved in the regulation of cells and their responses to diverse stimuli, however, the role of autophagy in glucocorticoid‑induced damage to bone marrow mesenchymal stem cells (BMSCs) remains unclear. The current study confirmed that glucocorticoid administration impaired the proliferation of BMSCs. Transmission electron microscopy, immunohistochemistry and western blot analysis detected autophagy in vitro and in GIOP model rats (in vivo). With the addition of the autophagy inhibitor 3‑methyladenine, the proliferative ability of BMSCs was further reduced, while the number of apoptotic BMSCs was significantly increased. The data suggests that in response to glucocorticoid administration, induced autophagy aids to maintain proliferation and prevent apoptosis of BMSCs. Thus, it is hypothesized that autophagy may be a novel target in the treatment or prevention of osteoporosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prednisolon, ≥99%
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Prednisolon, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glutaraldehyd -Lösung, technical, ~50% in H2O (5.6 M)