Merck
  • Value of 4D MR angiography at 3T compared with DSA for the follow-up of treated brain arteriovenous malformation.
Value of 4D MR angiography at 3T compared with DSA for the follow-up of treated brain arteriovenous malformation.
AJNR. American journal of neuroradiology (2014-06-07)
S Soize, F Bouquigny, K Kadziolka, C Portefaix, L Pierot
ZUSAMMENFASSUNG
Four-dimensional, contrast-enhanced MRA is a useful technique for the diagnosis and classification of brain AVM. The purpose of this study was to evaluate its usefulness in the follow-up of treated brain AVM. Patients with treated brain AVM (embolization, radiosurgery, and/or surgery) were investigated with both DSA (the "gold standard") and 4D MRA. Four-dimensional MRA was performed at 3T using a 4D sequence, combining contrast-enhanced timing-robust angiography, keyhole, and sensitivity encoding techniques. Examinations were evaluated by 2 independent readers and disagreements were resolved by a third reader. Interobserver and intermodality agreement with respect to residual nidus, residual venous drainage, and brain AVM patency were determined. Between May 2008 and February 2013, 37 patients with a median age of 45 years (interquartile range = 26-55) were prospectively included. Examinations were acquired 36 months (IQR = 10-45.5) after the last treatment. Interobserver agreement for brain AVM patency was very good for both 4D MRA (κ 0.82, 95% CI .67-.98) and DSA (κ 0.84, 95% CI .69-.98). After consensus reading, intermodality agreement for the evaluation of brain AVM patency was good (κ 0.73, 95% CI .55-.90). Diagnostic accuracy of 4D MRA for residual brain AVM compared with DSA, reached a sensitivity of 73.7%, specificity 100%, positive predictive value 100%, and negative predictive value 78.3%. Agreements by technique of treatment are also detailed. Four-dimensional MRA is a useful radiation-free technique for the follow-up of patients with treated brain AVM, especially patients treated by radiosurgery. However, given its actual limitations it is not sufficient to assert the cure; DSA remains mandatory for this purpose.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyl-D-Glucamin, 99.0-100.5% (titration)
Sigma-Aldrich
N-Methyl-D-Glucamin, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Meglumin, 99.0-100.5% dry basis, meets USP testing specifications
Supelco
Meglumin, Pharmaceutical Secondary Standard; Certified Reference Material
Iodixanol, European Pharmacopoeia (EP) Reference Standard
Meglumin, European Pharmacopoeia (EP) Reference Standard