Merck
TNF-alpha-308G>A polymorphism and the risk of familial CAD in a Pakistani population.
Human immunology (2014-12-17)
Sabir Hussain, Tahir Iqbal, Qamar Javed
ZUSAMMENFASSUNG
A case-control and trio-families study was performed to establish a potential association between TNF-alpha gene promoter SNPs at -308 and -238, and occurrence of CAD in a Pakistani population. In the first phase, 150 patients and 150 controls were enrolled in the case-control association study. In the second phase, heritability of susceptible alleles was investigated from 88 trio-families with CAD affected offspring. Biochemical analysis of lipids and hs-CRP was carried out spectrophotometrically, while serum TNF-alpha concentrations were determined by enzyme-linked immunosorbent assay. Genotyping of the TNF-alpha SNPs were determined by PCR-RFLP method. Elevated serum TNF-alpha and hs-CRP were observed from CAD vs. controls (P<0.0001; for both). The evaluation of TNF-alpha-308G>A polymorphism in case-control study revealed that the said SNP was significantly associated with the increased risk of CAD. The findings demonstrated a significant link between the TNF-alpha variant allele A at -308 and CAD (P=0.0035), whereas the -238 SNP was not associated with the disease. Haplotype A-G of the TNF-alpha gene at -308G>A and -238G>A showed higher frequency in the patient group compared with controls (P<0.05). Moreover, data showed preferential transmission of the disease susceptible allele A at TNF-alpha-308 from parent to affected offspring in a trio-family study (P<0.0001). The current research leads to conclusion that the TNF-alpha-308G>A polymorphism is associated with CAD in the study population. Furthermore, for the first time, we showed that the TNF-alpha-308A allele was significantly associated with the familial CAD in our high risk population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumtrichloracetat, 97%