Merck
  • Atorvastatin treatment improves the effects of mesenchymal stem cell transplantation on acute myocardial infarction: the role of the RhoA/ROCK/ERK pathway.
Atorvastatin treatment improves the effects of mesenchymal stem cell transplantation on acute myocardial infarction: the role of the RhoA/ROCK/ERK pathway.
International journal of cardiology (2014-08-21)
Qian Zhang, Hong Wang, Yue-Jin Yang, Qiu-Ting Dong, Tian-Jie Wang, Hai-Yan Qian, Na Li, Xi-Mei Wang, Chen Jin
ZUSAMMENFASSUNG
Statins protect mesenchymal stem cells (MSCs) against the harsh microenvironment and improve the efficacy of MSC transplantation after acute myocardial infarction (AMI); however, the mechanism remains uncertain. Furthermore, the transdifferentiation potential of MSCs in the post-infarct heart remains highly controversial. The RhoA/Rho-associated coiled-coil-forming kinase (ROCK) pathway participates in many aspects of the damaged heart after AMI and related to the "pleiotropic" effects of statins. This study aimed to explore whether atorvastatin (ATV) facilitates the survival and therapeutic efficacy of MSCs via the inhibition of RhoA/ROCK pathway and subsequently its downstream molecular extracellular regulated protein kinase (ERK1/2), and to investigate the transdifferentiation potential of MSCs in vivo. Female rats received myocardial injections of male rat MSCs 30 min after AMI. Four weeks after AMI, ATV combined with MSC treatment resulted in improved cardiac function and reduced infarct area. ATV facilitated the MSC survival, as revealed by the increased expression of Y chromosomal genes and the increased number of Y chromosome-positive cells; however, no transdifferentiation markers were observed. ATV inhibited the production of inflammatory cytokines both in vitro and vivo, accompanied by suppression of ROCK and ERK activities. Geranylgeranyl pyrophosphate (GGPP) abrogated the effects of ATV in the H9c2 cells under hypoxia/serum deprivation (H/SD), while the ROCK inhibitor fasudil mimicked the benefits of ATV after AMI. ATV improves the post-infarct microenvironment via RhoA/ROCK/ERK inhibition and thus facilitates the survival and efficacy of implanted MSCs. Transdifferentiation may be not responsible for the cardiac benefits that follow MSC transplantation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Chloral Hydrat, ≥99%
Sigma-Aldrich
Chloral Hydrat, crystallized, ≥98.0% (T)
Sigma-Aldrich
Geranylgeranylpyrophosphat Ammoniumsalz, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)