Merck
  • Sub-100nm gold nanomatryoshkas improve photo-thermal therapy efficacy in large and highly aggressive triple negative breast tumors.
Sub-100nm gold nanomatryoshkas improve photo-thermal therapy efficacy in large and highly aggressive triple negative breast tumors.
Journal of controlled release : official journal of the Controlled Release Society (2014-07-23)
Ciceron Ayala-Orozco, Cordula Urban, Sandra Bishnoi, Alexander Urban, Heather Charron, Tamika Mitchell, Martin Shea, Sarmistha Nanda, Rachel Schiff, Naomi Halas, Amit Joshi
ZUSAMMENFASSUNG
There is an unmet need for efficient near-infrared photothermal transducers for the treatment of highly aggressive cancers and large tumors where the penetration of light can be substantially reduced, and the intra-tumoral nanoparticle transport is restricted due to the presence of hypoxic or necrotic regions. We report the performance advantages obtained by sub 100nm gold nanomatryushkas, comprising concentric gold-silica-gold layers compared to conventional ~150nm silica core gold nanoshells for photothermal therapy of triple negative breast cancer. We demonstrate that a 33% reduction in silica-core-gold-shell nanoparticle size, while retaining near-infrared plasmon resonance, and keeping the nanoparticle surface charge constant, results in a four to five fold tumor accumulation of nanoparticles following equal dose of injected gold for both sizes. The survival time of mice bearing large (>1000mm(3)) and highly aggressive triple negative breast tumors is doubled for the nanomatryushka treatment group under identical photo-thermal therapy conditions. The higher absorption cross-section of a nanomatryoshka results in a higher efficiency of photonic to thermal energy conversion and coupled with 4-5× accumulation within large tumors results in superior therapy efficacy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems