Merck
  • Steroidal sapogenins and glycosides from the fibrous roots of Polygonatum odoratum with inhibitory effect on tissue factor (TF) procoagulant activity.
Steroidal sapogenins and glycosides from the fibrous roots of Polygonatum odoratum with inhibitory effect on tissue factor (TF) procoagulant activity.
Steroids (2014-07-22)
Hong Zhang, Ling Chen, Jun-Ping Kou, Dan-Ni Zhu, Jin Qi, Bo-Yang Yu
ZUSAMMENFASSUNG
Six new spirostane glycosides (1-6), named polygodosides A-F, one new furostanol glycoside, polygodoside G (7), one new cholestane glycoside, polygodoside H (8), and one new steroidal sapogenin, polygodosin A (9), together with thirteen known compounds (10-22) were isolated from a 90% MeOH extract of the fibrous roots of Polygonatum odoratum (Mill.) Druce. The structures of new compounds were elucidated by extensive 1D and 2D NMR spectroscopic analyses and mass spectrometry. The effects on TF procoagulant activity in THP-1 cells were tested for most of the compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Chlormethan -Lösung, 200 μg/mL in methanol, analytical standard
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chlormethan -Lösung, 1.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Chlormethan -Lösung, 1.0 M in diethyl ether