Merck
  • Novel 5-HT6 receptor antagonists/D2 receptor partial agonists targeting behavioral and psychological symptoms of dementia.
Novel 5-HT6 receptor antagonists/D2 receptor partial agonists targeting behavioral and psychological symptoms of dementia.
European journal of medicinal chemistry (2015-01-06)
Marcin Kołaczkowski, Monika Marcinkowska, Adam Bucki, Joanna Śniecikowska, Maciej Pawłowski, Grzegorz Kazek, Agata Siwek, Magdalena Jastrzębska-Więsek, Anna Partyka, Anna Wasik, Anna Wesołowska, Paweł Mierzejewski, Przemyslaw Bienkowski
ZUSAMMENFASSUNG
We describe a novel class of designed multiple ligands (DMLs) combining serotonin 5-HT6 receptor (5-HT6R) antagonism with dopamine D2 receptor (D2R) partial agonism. Prototype hybrid molecules were designed using docking to receptor homology models. Diverse pharmacophore moieties yielded 3 series of hybrids with varying in vitro properties at 5-HT6R and D2R, and at M1 receptor and hERG channel antitargets. 4-(piperazin-1-yl)-1H-indole derivatives showed highest antagonist potency at 5-HT6R, with 7-butoxy-3,4-dihydroquinolin-2(1H)-one and 2-propoxybenzamide derivatives having promising D2R partial agonism. 2-(3-(4-(1-(phenylsulfonyl)-1H-indol-4-yl)piperazin-1-yl)propoxy)benzamide (47) exhibited nanomolar affinity at both 5-HT6R and D2R and was evaluated in rat models. It displayed potent antidepressant-like and anxiolytic-like activity in the Porsolt and Vogel tests, respectively, more pronounced than that of a reference selective 5-HT6R antagonist or D2R partial agonist. In addition, 47 also showed antidepressant-like activity (Porsolt's test) and anxiolytic-like activity (open field test) in aged (>18-month old) rats. In operant conditioning tests, 47 enhanced responding for sweet reward in the saccharin self-administration test, consistent with anti-anhedonic properties. Further, 47 facilitated extinction of non-reinforced responding for sweet reward, suggesting potential procognitive activity. Taken together, these studies suggest that DMLs combining 5-HT6R antagonism and D2R partial agonism may successfully target affective disorders in patients from different age groups without a risk of cognitive deficits.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Saccharin, ≥99%
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143091, Saccharin, traceable to primary standards (LGC)
Sigma-Aldrich
Kaliumhydrid, in paraffin
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Saccharin, European Pharmacopoeia (EP) Reference Standard